Anti-MME Rabbit Polyclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB13114-100EA 2550 DKK
BIRBORB13114-100
Anti-MME Rabbit Polyclonal Antibody
Antistoffer
Western Blot: 1:500

Type: Primary
Antigen: MME (membrane metallo-endopeptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR