Anti-GMCSFR alpha Rabbit polyclonal antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB13109-100EA 2550 DKK
BIRBORB13109-100
Anti-GMCSFR alpha Rabbit polyclonal antibody
Antistoffer
Western Blot: 1:1000; Immunofluorescence: 1:250

Type: Primary
Antigen: GMCSFR alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR