Anti-MCM5 Goat Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB12980-100EA 0 DKK
BIRBORB12980-100
Anti-MCM5 Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MCM5 (minichromosome maintenance complex component 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Zebrafish
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR