Anti-ZIC3 Goat Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB12398-100EA 0 DKK
BIRBORB12398-100
Anti-ZIC3 Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ZIC3 (Zic family member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR