Anti-STK3 Goat Polyclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB12387-100EA 2710 DKK
BIRBORB12387-100
Anti-STK3 Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: STK3 (serine/threonine kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR