Anti-TBP Goat Polyclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB12374-100EA 2710 DKK
BIRBORB12374-100
Anti-TBP Goat Polyclonal Antibody
Antistoffer
ELISA: 1:64000; Western Blot: 0.3-1 ?g/ml

Type: Primary
Antigen: TBP (TATA box binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR