Anti-SFTPA1 Goat Polyclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB12372-100EA 2710 DKK
BIRBORB12372-100
Anti-SFTPA1 Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SFTPA1 (surfactant protein A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR