Anti-STRA6 Goat Polyclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB12360-100EA 2170 DKK
BIRBORB12360-100
Anti-STRA6 Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: STRA6 (stimulated by retinoic acid 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR