Anti-SLC25A38 Goat Polyclonal Antibody
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB12324-100EA 0 DKK
BIRBORB12324-100
Anti-SLC25A38 Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SLC25A38 (solute carrier family 25, member 38)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestil nu