Anti-LSM3 Goat Polyclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB12310-100EA 2170 DKK
BIRBORB12310-100
Anti-LSM3 Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: LSM3 (LSM3 homolog, U6 small nuclear RNA associated (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR