Anti-ABCD2 Goat Polyclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB12309-100EA 2710 DKK
BIRBORB12309-100
Anti-ABCD2 Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ABCD2 (ATP-binding cassette, sub-family D (ALD), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR