Anti-HA tag E. coli Monoclonal Antibody
Leverandør: Biorbyt
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB12253-100EA 2060 DKK
BIRBORB12253-100 BIRBORB12253-50
Anti-HA tag E. coli Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: HA tag (HA tag)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: E. coli
Isotype: IgG2a (Mouse)
Reactivity:
Bestil nu