Anti-IgG Rabbit Monoclonal Antibody (Biotin)

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB12248-100EA 1630 DKK
BIRBORB12248-100
Anti-IgG Rabbit Monoclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: IgG (IgG)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR