Anti-IgG1/IgG1 Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin)

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB12207-20EA 1630 DKK
BIRBORB12207-20 BIRBORB12207-50
Anti-IgG1/IgG1 Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin)
Antistoffer
Type: Primary
Antigen: IgG1/IgG1
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)/PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1+IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR