Anti-IgM Mouse Monoclonal Antibody (APC (Allophycocyanin))

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB12206-100EA 2060 DKK
BIRBORB12206-100 BIRBORB12206-50
Anti-IgM Mouse Monoclonal Antibody (APC (Allophycocyanin))
Antistoffer
Type: Primary
Antigen: IgM (IgM)
Clonality: monoclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR