Anti-IgG2b Mouse Monoclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB12197-500EA 2060 DKK
BIRBORB12197-500 BIRBORB12197-100
Anti-IgG2b Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: IgG2b
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR