Anti-IgG2a Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB12195-100EA 0 DKK
BIRBORB12195-100 BIRBORB12195-50
Anti-IgG2a Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®)
Antistoffer
Type: Primary
Antigen: IgG2a
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestil nu