Anti-CD4 Rat Monoclonal Antibody (APC (Allophycocyanin))

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB12178-50EA 1800 DKK
BIRBORB12178-50 BIRBORB12178-100
Anti-CD4 Rat Monoclonal Antibody (APC (Allophycocyanin))
Antistoffer
Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: monoclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR