Anti-CD235 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Leverandør: Biorbyt
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB12149-50EA 1800 DKK
BIRBORB12149-50 BIRBORB12149-100
Anti-CD235 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antistoffer
Type: Primary
Antigen: CD235
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human