Anti-CD235 Mouse Monoclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB12147-500EA 3030 DKK
BIRBORB12147-500 BIRBORB12147-100
Anti-CD235 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: CD235
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR