Anti-VCAM1 Mouse Monoclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB12135-500EA 3030 DKK
BIRBORB12135-500 BIRBORB12135-100
Anti-VCAM1 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: VCAM1 (vascular cell adhesion molecule 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR