Anti-CD44 Mouse Monoclonal Antibody (FITC (Fluorescein))

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB12058-100EA 2330 DKK
BIRBORB12058-100 BIRBORB12058-50
Anti-CD44 Mouse Monoclonal Antibody (FITC (Fluorescein))
Antistoffer
Type: Primary
Antigen: CD44 (CD44 molecule (Indian blood group))
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR