Anti-ITGA2B Mouse Monoclonal Antibody (PE (Phycoerythrin))

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB12053-100EA 2440 DKK
BIRBORB12053-100 BIRBORB12053-50
Anti-ITGA2B Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antistoffer
FACS: 1:50-100

Type: Primary
Antigen: ITGA2B (integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41))
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR