Anti-CD38 Mouse Monoclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB12043-100EA 1630 DKK
BIRBORB12043-100 BIRBORB12043-500
Anti-CD38 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: CD38 (CD38 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR