Anti-MS4A1 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®)

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB12000-50EA 2010 DKK
BIRBORB12000-50 BIRBORB12000-100
Anti-MS4A1 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®)
Antistoffer
Type: Primary
Antigen: MS4A1 (membrane-spanning 4-domains, subfamily A, member 1)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR