Anti-CD5 Mouse Monoclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB11940-500EA 3030 DKK
BIRBORB11940-500 BIRBORB11940-100
Anti-CD5 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: CD5 (CD5 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR