Anti-CD3 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®)

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB11924-50EA 2010 DKK
BIRBORB11924-50 BIRBORB11924-100
Anti-CD3 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®)
Antistoffer
Type: Primary
Antigen: CD3
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR