Anti-CD3 Mouse Monoclonal Antibody (FITC (Fluorescein))
Leverandør: Biorbyt
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB11922-50EA 1800 DKK
BIRBORB11922-50 BIRBORB11922-100
Anti-CD3 Mouse Monoclonal Antibody (FITC (Fluorescein))
Antistoffer
Type: Primary
Antigen: CD3
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human