Anti-RAB28 Goat Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB11632-100EA 0 DKK
BIRBORB11632-100
Anti-RAB28 Goat Polyclonal Antibody
Antistoffer
Western Blot: 1:250-1:2000

Type: Primary
Antigen: RAB28 (RAB28, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR