Anti-EEA1 Goat polyclonal antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB11606-100EA 1800 DKK
BIRBORB11606-100
Anti-EEA1 Goat polyclonal antibody
Antistoffer
Western Blot: 1:250-1:1000

Type: Primary
Antigen: EEA1 (early endosome antigen 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR