Anti-YY1 Rabbit Polyclonal Antibody
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB11577-100EA 0 DKK
BIRBORB11577-100 BIRBORB11577-200
Anti-YY1 Rabbit Polyclonal Antibody
Antistoffer
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: YY1 (YY1 transcription factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat