Anti-SSTR1 Rabbit Polyclonal Antibody
Leverandør: Biorbyt
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB11421-100EA 2330 DKK
BIRBORB11421-100 BIRBORB11421-200
Anti-SSTR1 Rabbit Polyclonal Antibody
Antistoffer
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: SSTR1 (somatostatin receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat