Anti-ITGA5 Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB10945-100EA 0 DKK
BIRBORB10945-100 BIRBORB10945-200
Anti-ITGA5 Rabbit Polyclonal Antibody
Antistoffer
Western Blot: 2g/ml; Immunochemistry: 1:500; Immunofluorescence: 1:500

Type: Primary
Antigen: ITGA5 (integrin, alpha 5 (fibronectin receptor, alpha polypeptide))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR