Anti-INS Rabbit Polyclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB10924-100EA 2500 DKK
BIRBORB10924-100 BIRBORB10924-200
Anti-INS Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: INS (insulin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR