Anti-BRI3BP Rabbit polyclonal antibody
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB10781-100EA 0 DKK
BIRBORB10781-100 BIRBORB10781-200
Anti-BRI3BP Rabbit polyclonal antibody
Antistoffer
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: BRI3BP (BRI3 binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human