Anti-PMP2 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB9313-100EA 2880 DKK
BIRBORB9313-100
Anti-PMP2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: PMP2 (peripheral myelin protein 2)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR