Anti-PTK2B Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB8880-100EA 2880 DKK
BIRBORB8880-100
Anti-PTK2B Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: PTK2B (protein tyrosine kinase 2 beta)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR