Anti-AGPAT2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB8707-100EA 0 DKK
BIRBORB8707-100
Anti-AGPAT2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: AGPAT2 (1-acylglycerol-3-phosphate O-acyltransferase 2)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human