Anti-MDA-LDL Mouse Monoclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB8259-100EA 4980 DKK
BIRBORB8259-100
Anti-MDA-LDL Mouse Monoclonal Antibody
Antistoffer
Western Blot: 1:1000-1:3000

Type: Primary
Antigen: MDA-LDL
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Murine)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR