Anti-HLA-DR Mouse Monoclonal Antibody (PE (Phycoerythrin))

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB8225-100EA 4060 DKK
BIRBORB8225-100
Anti-HLA-DR Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antistoffer
Western Blot: 1:1000-1:3000

Type: Primary
Antigen: HLA-DR
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR