Anti-FCGR3A Mouse Monoclonal Antibody (PE (Phycoerythrin))

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB8134-100EA 4980 DKK
BIRBORB8134-100
Anti-FCGR3A Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antistoffer
Western Blot: 1:1000-1:3000

Type: Primary
Antigen: FCGR3A (Fc fragment of IgG, low affinity IIIa, receptor (CD16a))
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR