Anti-Fibrinogen Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB8094-100EA 0 DKK
BIRBORB8094-100
Anti-Fibrinogen Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: Fibrinogen
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR