Anti-JMJD7 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB7699-100EA 2880 DKK
BIRBORB7699-100
Anti-JMJD7 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: JMJD7 (jumonji domain containing 7)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR