Anti-TSPAN6 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB7187-100EA 0 DKK
BIRBORB7187-100
Anti-TSPAN6 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: TSPAN6 (tetraspanin 6)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR