Anti-SLC9A9 Rabbit Polyclonal Antibody
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB6962-100EA 0 DKK
BIRBORB6962-100 BIRBORB6962-200
Anti-SLC9A9 Rabbit Polyclonal Antibody
Antistoffer
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: SLC9A9 (solute carrier family 9, subfamily A (NHE9, cation proton antiporter 9), member 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat