Anti-AKT3 Rabbit Polyclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB6787-100EA 2330 DKK
BIRBORB6787-100 BIRBORB6787-200
Anti-AKT3 Rabbit Polyclonal Antibody
Antistoffer
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: AKT3 (v-akt murine thymoma viral oncogene homolog 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR