Anti-ICAM1 Rabbit Polyclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB6189-20EA 1650 DKK
BIRBORB6189-20 BIRBORB6189-100
Anti-ICAM1 Rabbit Polyclonal Antibody
Antistoffer
Western Blot: 1:500-1:2000

Type: Primary
Antigen: ICAM1 (intercellular adhesion molecule 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR