Anti-PLC gamma 2 Rabbit Polyclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB6042-100EA 2280 DKK
BIRBORB6042-100
Anti-PLC gamma 2 Rabbit Polyclonal Antibody
Antistoffer
Immunochemistry: 1:50-100

Type: Primary
Antigen: PLC gamma 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR