Anti-SERPINB1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Leverandør: Biorbyt
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB5318-100EA 2880 DKK
BIRBORB5318-100
Anti-SERPINB1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)
Type: Primary
Antigen: SERPINB1 (serpin peptidase inhibitor, clade B (ovalbumin), member 1)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat