Anti-MRC1 Rabbit Polyclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB4941-200EA 3790 DKK
BIRBORB4941-200 BIRBORB4941-100
Anti-MRC1 Rabbit Polyclonal Antibody
Antistoffer
Western Blot: 200: 1500; Immunochemistry: 1:100-500

Type: Primary
Antigen: MRC1 (mannose receptor, C type 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR